accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 43210-67-9

H3COOHNHNSN

CAS RN 43210-67-9

IUPAC Name: methyl N-[6-(phenylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: fenbendazole
Molecular Weight (g/mol): 299.35
Molecular Formula: C15H13N3O2S
InChi Key: HDDSHPAODJUKPD-UHFFFAOYSA-N
SMILES: COC(=O)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results