accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 426831-32-5

H3COFOH

CAS RN 426831-32-5

IUPAC Name: (5-fluoro-2-methoxyphenyl)methanol
Synonyms: (5-fluoro-2-methoxyphenyl)methanol
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: NJDNHGOIOZXFCB-UHFFFAOYSA-N
SMILES: COC1=C(CO)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results