accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNHSOOCH3(E/Z)

CAS RN 41780-81-8

IUPAC Name: 4-methyl-N'-(1-phenylbutan-2-ylidene)benzene-1-sulfonohydrazide
Synonyms: 4-methyl-N'-(1-phenylbutan-2-ylidene)benzenesulfonohydrazide
Molecular Weight (g/mol): 316.42
Molecular Formula: C17H20N2O2S
InChi Key: DQAAEKVIAXFJEK-UHFFFAOYSA-N
SMILES: CCC(CC1=CC=CC=C1)=NNS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results