CAS RN 4152-09-4
CAS RN 4152-09-4
IUPAC Name:
(2-aminoethyl)(benzyl)amine
Synonyms:
(2-aminoethyl)(benzyl)amine
Molecular Weight (g/mol):
150.23
Molecular Formula:
C9H14N2
InChi Key:
ACYBVNYNIZTUIL-UHFFFAOYSA-N
SMILES:
NCCNCC1=CC=CC=C1
1
–
3
of
3
results
1
N-Benzylethylenediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Benzylethylenediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Benzylethylenediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.