Learn More
N-Benzylethylenediamine, TRC
Specifications
Specifications
| Chemical Name or Material | Monobenzylethylendiamine |
| CAS | 4152-09-4 |
| Molecular Formula | C9 H14 N2 |
| Quantity | 10 g |
| Synonym | Monobenzylethylendiamine, Benzylpenicillin Benzathine Imp. A (EP) |
| InChI Formula | InChI=1S/C9H14N2/c10-6-7-11-8-9-4-2-1-3-5-9/h1-5,11H,6-8,10H2 |
| SMILES | NCCNCc1ccccc1 |
| IUPAC Name | N'-benzylethane-1,2-diamine |
| Molecular Weight (g/mol) | 150.22 |
| Formula Weight | 150.1157 |