accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOH2COOCH3

CAS RN 40637-56-7

IUPAC Name: 1,3-dimethyl 2-(prop-2-en-1-yl)propanedioate
Synonyms: 1,3-dimethyl 2-(prop-2-en-1-yl)propanedioate
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: VZNFVLWVVHHMBG-UHFFFAOYSA-N
SMILES: COC(=O)C(CC=C)C(=O)OC
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results