Learn More
Dimethyl Allylmalonate, TRC
Specifications
Specifications
| Chemical Name or Material | Dimethyl Allylmalonate |
| CAS | 40637-56-7 |
| Molecular Formula | C8H12O4 |
| Quantity | 1 g |
| Synonym | Dimethyl 2-Allylmalonate, Dimethyl 2-(Prop-2-enyl)propane-1,3-dioate, 4,4-Bis(carboxymethoxy)-1-butene, Dimethyl 2-(2-propenyl)malonate, 2-propenylpropanedioic Acid Dimethyl Ester |
| InChI Formula | InChI=1S/C8H12O4/c1-4-5-6(7(9)11-2)8(10)12-3/h4,6H,1,5H2,2-3H3 |
| SMILES | COC(C(C(OC)=O)CC=C)=O |
| IUPAC Name | dimethyl 2-allylmalonate |
| Molecular Weight (g/mol) | 172.18 |
| Formula Weight | 172.07 |