accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 405-04-9

NFOH

CAS RN 405-04-9

IUPAC Name: 3-fluoro-4-hydroxybenzonitrile
Synonyms: 3-fluoro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 137.11
Molecular Formula: C7H4FNO
InChi Key: DPSSSDFTLVUJDH-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results