CAS RN 398473-34-2
IUPAC Name:
1-(3-{4-[(piperidin-1-yl)methyl]phenoxy}propyl)piperidine
Synonyms:
1-{3-[4-(piperidin-1-ylmethyl)phenoxy]propyl}piperidine
Molecular Weight (g/mol):
316.49
Molecular Formula:
C20H32N2O
InChi Key:
PTKHFRNHJULJKT-UHFFFAOYSA-N
SMILES:
C(COC1=CC=C(CN2CCCCC2)C=C1)CN1CCCCC1
1
–
2
of
2
results
1
Tocris Bioscience™ JNJ 5207852 dihydrochloride
CAS: 398473-34-2 Molecular Formula: C20H32N2O
2
JNJ-5207852, MedChemExpress
MedChemExpress JNJ-5207852 is a selective and potent histamine H3 receptor (H3R) antagonist, with pKis of 8.9, 9.24 for rat and human H3R, respectively.