Learn More
JNJ-5207852, MedChemExpress
Product Code. 16485008
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485008 10 mg 10mg
16481247 50 mg 50mg
16467737 5 mg 5mg
16455766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16485008 Supplier Medchem Express Supplier No. HY12190
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
MedChemExpress JNJ-5207852 is a selective and potent histamine H3 receptor (H3R) antagonist, with pKis of 8.9, 9.24 for rat and human H3R, respectively.
JNJ-5207852 is a selective and potent histamine H3 receptor (H3R) antagonist, with pKis of 8.9, 9.24 for rat and human H3R, respectively.
Specifications
Chemical Name or Material JNJ-5207852
CAS 398473-34-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H32N2O
Quantity 10 mg
Solubility Information DMSO : 77.5 mg/mL (244.88 mM; Need ultrasonic)
SMILES N1(CCCOC2=CC=C(CN3CCCCC3)C=C2)CCCCC1
Molecular Weight (g/mol) 316.48
Formula Weight 316.48
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation