accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3970-42-1

IUPAC Name: 1-(2-chloro-6-nitrophenyl)piperidine
Synonyms: 1-(2-chloro-6-nitrophenyl)piperidine
Molecular Weight (g/mol): 240.69
Molecular Formula: C11H13ClN2O2
InChi Key: IBRSUFPYKBRXMU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(N2CCCCC2)C(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results