accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3964-56-5

OHBrCl

CAS RN 3964-56-5

IUPAC Name: 4-bromo-2-chlorophenol
Synonyms: 4-bromo-2-chlorophenol C6H4BrClO C6-H4-br-cl-O
Molecular Weight (g/mol): 207.45
Molecular Formula: C6H4BrClO
InChi Key: VIBJPUXLAKVICD-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1Cl
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)