CAS RN 3964-52-1
CAS RN 3964-52-1
IUPAC Name:
4-amino-2-chlorophenol
Synonyms:
4-amino-2-chlorophenol
Molecular Weight (g/mol):
143.57
Molecular Formula:
C6H6ClNO
InChi Key:
ZYZQSCWSPFLAFM-UHFFFAOYSA-N
SMILES:
NC1=CC=C(O)C(Cl)=C1
1
–
1
of
1
results
1
4-Amino-2-chlorophenol, 99%
CAS: 3964-52-1 Molecular Formula: C6H6ClNO Molecular Weight (g/mol): 143.57 MDL Number: MFCD00128891 InChI Key: ZYZQSCWSPFLAFM-UHFFFAOYSA-N Synonym: 3-chloro-4-hydroxyaniline,2-chloro-4-aminophenol,phenol, 4-amino-2-chloro,4-amino-2-chloro-phenol,unii-2z6zt3b98u,o-chloro-p-aminophenol,pubchem18907,salzsaurem p-aminophenol,acmc-1aiqv,3-chloro4-hydroxyaniline PubChem CID: 77578 IUPAC Name: 4-amino-2-chlorophenol SMILES: NC1=CC=C(O)C(Cl)=C1