CAS RN 383134-85-8
CAS RN 383134-85-8
IUPAC Name:
2-(5-fluoro-2-methoxyphenyl)acetic acid
Synonyms:
(5-fluoro-2-methoxyphenyl)acetic acid
Molecular Weight (g/mol):
184.17
Molecular Formula:
C9H9FO3
InChi Key:
WGZCWKASVDIAOH-UHFFFAOYSA-N
SMILES:
COC1=C(CC(O)=O)C=C(F)C=C1
1
–
1
of
1
results
1
5-Fluoro-2-methoxyphenylacetic acid, 97%, Thermo Scientific™
CAS: 383134-85-8 Molecular Formula: C9H9FO3 Molecular Weight (g/mol): 184.17 MDL Number: MFCD00671771 InChI Key: WGZCWKASVDIAOH-UHFFFAOYSA-N Synonym: 2-5-fluoro-2-methoxyphenyl acetic acid,5-fluoro-2-methoxyphenylacetic acid,5-fluoro-2-methoxybenzeneacetic acid,5-fluoro-2-methoxyphenyl acetic acid,2-methoxy-5-fluorophenylacetic acid,rarechem al ms 0116,5-fluoro-2-methoxy-phenyl-acetic acid PubChem CID: 302684 IUPAC Name: 2-(5-fluoro-2-methoxyphenyl)acetic acid SMILES: COC1=C(CC(O)=O)C=C(F)C=C1