CAS RN 37519-14-5
CAS RN 37519-14-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acetaminophen metabolite 3-hydroxy-acetaminophen, MedChemExpress
MedChemExpress 3-hydroxy-acetaminophen is a metabolite of Acetaminophen, which is a pain medicine.