accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3680-02-2

CH3SOOH2C

CAS RN 3680-02-2

IUPAC Name: methanesulfonylethene
Synonyms: methyl vinyl sulfone
Molecular Weight (g/mol): 106.14
Molecular Formula: C3H6O2S
InChi Key: WUIJTQZXUURFQU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C=C
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)