accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3663-80-7

OO(S)OO

CAS RN 3663-80-7

IUPAC Name: (2S)-2,3-dihydro-1,4-benzodioxine-2-carboxylate
Synonyms: (2S)-2,3-dihydro-1,4-benzodioxine-2-carboxylate
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H7O4
InChi Key: HMBHAQMOBKLWRX-QMMMGPOBSA-M
SMILES: [O-]C(=O)[C@@H]1COC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results