accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3NH2NN

CAS RN 36315-01-2

IUPAC Name: 4,6-dimethoxypyrimidin-2-amine
Synonyms: 4,6-dimethoxypyrimidin-2-amine
Molecular Weight (g/mol): 155.16
Molecular Formula: C6H9N3O2
InChi Key: LVFRCHIUUKWBLR-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(N)=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)