Learn More
2-Amino-4,6-dimethoxypyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4,6-dimethoxypyrimidine |
| CAS | 36315-01-2 |
| Molecular Formula | C6 H9 N3 O2 |
| Quantity | 1 g |
| Synonym | Pyrimidine, 2-amino-4,6-dimethoxy- (7CI), 4,6-Dimethoxy-2-pyrimidinamine, 2-Amino-4,6-dimethoxypyrimidine, 4,6-Dimethoxy-2-aminopyrimidine, AE-F 092944, J 290, ADPM, IN-J0290 |
| InChI Formula | InChI=1S/C6H9N3O2/c1-10-4-3-5(11-2)9-6(7)8-4/h3H,1-2H3,(H2,7,8,9) |
| SMILES | COc1cc(OC)nc(N)n1 |
| IUPAC Name | 4,6-dimethoxypyrimidin-2-amine |
| Molecular Weight (g/mol) | 155.15 |
| Formula Weight | 155.0695 |