accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SH3C

CAS RN 352-93-2

IUPAC Name: (ethylsulfanyl)ethane
Synonyms: ethyl sulfide sulfodor thioethyl ether diethylsulfid
Molecular Weight (g/mol): 90.18
Molecular Formula: C4H10S
InChi Key: LJSQFQKUNVCTIA-UHFFFAOYSA-N
SMILES: CCSCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)