accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFSOOCl

CAS RN 351003-43-5

IUPAC Name: 3,4,5-trifluorobenzene-1-sulfonyl chloride
Synonyms: 3,4,5-trifluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 230.59
Molecular Formula: C6H2ClF3O2S
InChi Key: HSEHKULCGSMHAE-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1F)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results