accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SOOClNF

CAS RN 351003-23-1

IUPAC Name: 3-cyano-4-fluorobenzene-1-sulfonyl chloride
Synonyms: 3-cyano-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 219.61
Molecular Formula: C7H3ClFNO2S
InChi Key: KDVZADPGGPBAAK-UHFFFAOYSA-N
SMILES: FC1=C(C=C(C=C1)S(Cl)(=O)=O)C#N
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)