accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClSClOO

CAS RN 34981-38-9

IUPAC Name: 5-chloro-2-methylbenzene-1-sulfonyl chloride
Synonyms: 5-chloro-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: VHBFBNCERXCECQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results