accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHOFFF

CAS RN 349-56-4

IUPAC Name: 3-methoxy-5-(trifluoromethyl)phenol
Synonyms: 3-methoxy-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 192.14
Molecular Formula: C8H7F3O2
InChi Key: DFOFNISQFZKBEY-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results