accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 34420-17-2

HOBOH

CAS RN 34420-17-2

IUPAC Name: (2-phenylethyl)boronic acid
Synonyms: phenylethane boronic acid PEBA
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: VPRUMANMDWQMNF-UHFFFAOYSA-N
SMILES: OB(O)CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results