accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 34145-05-6

HOClCl

CAS RN 34145-05-6

IUPAC Name: (2,5-dichlorophenyl)methanol
Synonyms: (2,5-dichlorophenyl)methanol
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: LCEIGNVIDJNUGF-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=CC=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results