accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BH3CCH3OHOH

CAS RN 333432-28-3

IUPAC Name: (9,9-dimethyl-9H-fluoren-2-yl)boronic acid
Synonyms: 9,9-dimethylfluoren-2-ylboronic acid
Molecular Weight (g/mol): 238.09
Molecular Formula: C15H15BO2
InChi Key: DMDPAJOXRYGXCB-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results