accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNHClNOO

CAS RN 332023-12-8

IUPAC Name: 1-(2-chloro-6-nitrophenyl)piperazine
Synonyms: 1-(2-chloro-6-nitrophenyl)piperazine
Molecular Weight (g/mol): 241.68
Molecular Formula: C10H12ClN3O2
InChi Key: GKOXZZHKOZLHRQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(N2CCNCC2)C(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results