CAS RN 331943-04-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-[3-(4-Methylphenyl)-5-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-De(4-sulfonamidophenyl)-N'-(4-sulfonamidophenyl) Celecoxib, TRC
CAS: 331943-04-5 Molecular Formula: C17 H14 F3 N3 O2 S Molecular Weight (g/mol): 381.37 Synonym: Benzenesulfonamide, 4-[3-(4-methylphenyl)-5-(trifluoromethyl)-1H-pyrazol-1-yl]-,Celecoxib Imp. B (EP),Celecoxib RC B (USP) IUPAC Name: 4-[3-(4-methylphenyl)-5-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide SMILES: Cc1ccc(cc1)c2cc(n(n2)c3ccc(cc3)S(=O)(=O)N)C(F)(F)F