CAS RN 330942-05-7
CAS RN 330942-05-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Betrixaban, MedChemExpress
MedChemExpress Betrixaban (PRT054021) is a highly potent, selective, and orally efficacious factor Xa (fXa) inhibitor with an IC50 of 1.5 nM. Betrixaban shows antithrombotic effect.