Learn More
Betrixaban, MedChemExpress
Product Code. 16486477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16486477
Brand: Medchem Express HY102685mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Betrixaban (PRT054021) is a highly potent, selective, and orally efficacious factor Xa (fXa) inhibitor with an IC50 of 1.5 nM. Betrixaban shows antithrombotic effect.
Betrixaban (PRT054021) is a highly potent, selective, and orally efficacious factor Xa (fXa) inhibitor with an IC50 of 1.5 nM. Betrixaban shows antithrombotic effect.
Specifications
Chemical Name or Material Betrixaban
CAS 330942-05-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H22ClN5O3
Quantity 5 mg
Synonym PRT054021
Solubility Information DMSO : 25 mg/mL (55.32 mM; Need ultrasonic)
SMILES N=C(N(C)C)C(C=C1)=CC=C1C(NC2=CC=C(OC)C=C2C(NC(C=C3)=NC=C3Cl)=O)=O
Molecular Weight (g/mol) 451.91
Formula Weight 451.91
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation