CAS RN 330203-01-5
CAS RN 330203-01-5
IUPAC Name:
5-[(3-bromo-4-hydroxyphenyl)methylidene]-3-(4-methoxyphenyl)-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms:
5-[(3-bromo-4-hydroxyphenyl)methylidene]-3-(4-methoxyphenyl)-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol):
422.31
Molecular Formula:
C17H12BrNO3S2
InChi Key:
HEFKAUYVLSGPGC-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)N1C(=S)SC(=CC2=CC(Br)=C(O)C=C2)C1=O
1
–
1
of
1
results
1
TC HSD 21, MedChemExpress
MedChemExpress TC HSD 21 is a potent 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) inhibitor with an IC50 of 14 nM. TC HSD 21 shows excellent selectivity over 17β-HSD isoenzymes and nuclear receptors.