Learn More
TC HSD 21, MedChemExpress
Product Code. 16426567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16426567 Supplier Medchem Express Supplier No. HY103394
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
MedChemExpress TC HSD 21 is a potent 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) inhibitor with an IC50 of 14 nM. TC HSD 21 shows excellent selectivity over 17β-HSD isoenzymes and nuclear receptors.
TC HSD 21 is a potent 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) inhibitor with an IC50 of 14 nM. TC HSD 21 shows excellent selectivity over 17β-HSD isoenzymes and nuclear receptors.
Specifications
Chemical Name or Material TC HSD 21
CAS 330203-01-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H12BrNO3S2
Quantity 5 mg
SMILES O=C1N(C2=CC=C(OC)C=C2)C(S/C1=C\C3=CC=C(O)C(Br)=C3)=S
Molecular Weight (g/mol) 422.32
Formula Weight 422.32
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation