Learn More
2,4,5-Tribromotoluene, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,5-Tribromotoluene |
| CAS | 3278-88-4 |
| Molecular Formula | C7H5Br3 |
| Quantity | 500 mg |
| Synonym | Benzene','1,2,4-tribromo-5-methyl- (9CI','ACI), 1,2,4-Tribromo-5-methylbenzene (ACI), Toluene','2,4,5-tribromo- (6CI','7CI','8CI), 2,4,5-Tribromotoluene |
| InChI Formula | InChI=1S/C7H5Br3/c1-4-2-6(9)7(10)3-5(4)8/h2-3H,1H3 |
| SMILES | CC1=CC(=C(C=C1Br)Br)Br |
| IUPAC Name | 1,2,4-tribromo-5-methylbenzene |
| Molecular Weight (g/mol) | 328.82 |
| Formula Weight | 325.7941 |