CAS RN 32774-16-6
CAS RN 32774-16-6
IUPAC Name:
3,3',4,4',5,5'-hexachloro-1,1'-biphenyl
Synonyms:
3,3',4,4',5,5'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol):
360.86
Molecular Formula:
C12H4Cl6
InChi Key:
ZHLICBPIXDOFFG-UHFFFAOYSA-N
SMILES:
ClC1=CC(=CC(Cl)=C1Cl)C1=CC(Cl)=C(Cl)C(Cl)=C1
1
–
2
of
2
results
1
3,3',4,4',5,5'-Hexachlorobiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,3',4,4',5,5'-Hexachlorobiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.