Learn More
3,3',4,4',5,5'-Hexachlorobiphenyl, TRC
Specifications
Specifications
| Chemical Name or Material | PCB No. 169 |
| CAS | 32774-16-6 |
| Molecular Formula | C12 H4 Cl6 |
| Quantity | 250 mg |
| Synonym | 1,1'-Biphenyl, 3,3',4,4',5,5'-hexachloro-, Biphenyl, 3,3',4,4',5,5'-hexachloro- (8CI), 3,3',4,4',5,5'-Hexachloro-1,1'-biphenyl, 3,3',4,4',5,5'-Hexachlorobiphenyl, 3,4,5,3',4',5'-Hexachlorobiphenyl, CB 169, HxCB 169, PCB 169, 3,3',4,4',5,5'-HexaCB |
| InChI Formula | InChI=1S/C12H4Cl6/c13-7-1-5(2-8(14)11(7)17)6-3-9(15)12(18)10(16)4-6/h1-4H |
| SMILES | Clc1cc(cc(Cl)c1Cl)c2cc(Cl)c(Cl)c(Cl)c2 |
| IUPAC Name | 1,2,3-trichloro-5-(3,4,5-trichlorophenyl)benzene |
| Molecular Weight (g/mol) | 360.88 |
| Formula Weight | 357.8444 |