CAS RN 327048-99-7
CAS RN 327048-99-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Des-(Cyclohexa-2,5-dien-1-one) Boldenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Des-(Cyclohexa-2,5-dien-1-one) Boldenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Des-(Cyclohexa-2,5-dien-1-one) Boldenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.