Learn More
Des-(Cyclohexa-2,5-dien-1-one) Boldenone, TRC
Product Code. 30387271
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387271 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30387271 Supplier Toronto Research Chemicals Supplier No. TRCD6751305MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Des-(Cyclohexa-2,5-dien-1-one) Boldenone
CAS 327048-99-7
Molecular Formula C16H22O3
Quantity 5 mg
Synonym rac-(3aR,5aS,6R,9aS,9bS)-3a,6-Dimethyl-3,7-dioxododecahydro-1H-cyclopenta[a]naphthalene-6-carbaldehyde
InChI Formula InChI=1S/C16H22O3/c1-15-8-7-12-10(11(15)4-6-13(15)18)3-5-14(19)16(12,2)9-17/h9-12H,3-8H2,1-2H3/t10-,11-,12-,15-,16+/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC(=O)[C@]3(C)C=O)[C@@H]1CCC2=O
IUPAC Name (3aS,5aS,6S,9aS,9bS)-3a,6-dimethyl-3,7-dioxo-2,4,5,5a,8,9,9a,9b-octahydro-1H-cyclopenta[a]naphthalene-6-carbaldehyde
Molecular Weight (g/mol) 262.35
Formula Weight 262.1569