accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 327036-89-5

CH3NONOS

CAS RN 327036-89-5

IUPAC Name: 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Synonyms: 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Molecular Weight (g/mol): 222.26
Molecular Formula: C10H10N2O2S
InChi Key: JDSJDASOXWCHPN-UHFFFAOYSA-N
SMILES: CN1SC(=O)N(CC2=CC=CC=C2)C1=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)