CAS RN 327-98-0
CAS RN 327-98-0
IUPAC Name:
O-ethyl O-2,4,5-trichlorophenyl ethylphosphonothioate
Synonyms:
trichloronate
Molecular Weight (g/mol):
333.59
Molecular Formula:
C10H12Cl3O2PS
InChi Key:
ANIAQSUBRGXWLS-UHFFFAOYNA-N
SMILES:
CCOP(=S)(CC)OC1=CC(Cl)=C(Cl)C=C1Cl
1
–
2
of
2
results
1
Trichloronate, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality