CAS RN 32690-93-0
CAS RN 32690-93-0
IUPAC Name:
2,4,4',5-tetrachloro-1,1'-biphenyl
Synonyms:
2,4,4',5-tetrachlorobiphenyl
Molecular Weight (g/mol):
291.98
Molecular Formula:
C12H6Cl4
InChi Key:
TULCXSBAPHCWCF-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1=CC(Cl)=C(Cl)C=C1Cl
1
–
3
of
3
results
1
2,4,4',5-Tetrachlorobiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,4,4',5-Tetrachlorobiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,4,4',5-Tetrachlorobiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.