Learn More
2,4,4',5-Tetrachlorobiphenyl, TRC
Specifications
Specifications
| Chemical Name or Material | PCB No. 74 |
| CAS | 32690-93-0 |
| Molecular Formula | C12 H6 Cl4 |
| Quantity | 250 mg |
| Synonym | 1,1'-Biphenyl','2,4,4',5-tetrachloro-, Biphenyl','2,4,4',5-tetrachloro- (8CI), 2,4,4',5-Tetrachloro-1,1'-biphenyl, 2,4,4',5-Tetrachlorobiphenyl, 2,4,5,4'-Tetrachlorobiphenyl, PCB 74, 2,4,4',5-TetraCB |
| InChI Formula | InChI=1S/C12H6Cl4/c13-8-3-1-7(2-4-8)9-5-11(15)12(16)6-10(9)14/h1-6H |
| SMILES | Clc1ccc(cc1)c2cc(Cl)c(Cl)cc2Cl |
| IUPAC Name | 1,2,4-trichloro-5-(4-chlorophenyl)benzene |
| Molecular Weight (g/mol) | 291.99 |
| Formula Weight | 289.9224 |