CAS RN 326491-79-6
CAS RN 326491-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R)-1-(3,4-Dimethoxyphenyl)-1,2-ethanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.