CAS RN 325457-99-6
CAS RN 325457-99-6
IUPAC Name:
4-chloro-N-(6-chloropyridin-3-yl)benzamide
Synonyms:
4-chloro-N-(6-chloropyridin-3-yl)benzamide
Molecular Weight (g/mol):
267.11
Molecular Formula:
C12H8Cl2N2O
InChi Key:
FOYOGIUAMYSVIT-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C(=O)NC1=CN=C(Cl)C=C1
1
–
1
of
1
results
1