accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3252-36-6

H3COOHNBrCl

CAS RN 3252-36-6

IUPAC Name: 5-bromo-4-chloro-1H-indol-3-yl acetate
Synonyms: 5-bromo-4-chloro-1H-indol-3-yl acetate
Molecular Weight (g/mol): 288.53
Molecular Formula: C10H7BrClNO2
InChi Key: WPWLFFMSSOAORQ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)