Learn More
MedChemExpress BCDA
Specifications
Specifications
| CAS | 3252-36-6 |
| Solubility | Ethanol : ≥ 100 mg/mL (346.58 mM) |
| Synonym | 5-bromo-4-chloroindoxyl acetate |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 288.53 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 99.0% |
| Shipping Condition | Room temperature |