CAS RN 32509-66-3
CAS RN 32509-66-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Hostanox 03, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ethylene Bis[3,3-bis[3-tert-butyl-4-hydroxyphenyl]butanoate], Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Hostanox 03, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Hostanox 03, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.