CAS RN 324763-46-4
CAS RN 324763-46-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5(S)-[1(S)-Azido-3(S)-[4-methoxy-3-(3-methoxypropoxy)benzyl]-4-methylpentyl]-3(S)-isopropyldihydrofuran-2-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5(S)-[1(S)-Azido-3(S)-[4-methoxy-3-(3-methoxypropoxy)benzyl]-4-methylpentyl]-3(S)-isopropyldihydrofuran-2-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.