Learn More
5(S)-[1(S)-Azido-3(S)-[4-methoxy-3-(3-methoxypropoxy)benzyl]-4-methylpentyl]-3(S)-isopropyldihydrofuran-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 5(S)-[1(S)-Azido-3(S)-[4-methoxy-3-(3-methoxypropoxy)benzyl]-4-methylpentyl]-3(S)-isopropyldihydrofuran-2-one |
| CAS | 324763-46-4 |
| Molecular Formula | C25H39N3O5 |
| Quantity | 250 mg |
| Synonym | (3S,5S)-5-[(1S,3S)-1-Azido-3-[[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl]-4-methylpentyl]dihydro-3-(1-methylethyl)-2(3H)-furanone |
| InChI Formula | InChI=1S/C25H39N3O4/c1-16(2)20(13-18-9-10-23(31-6)19(12-18)8-7-11-30-5)14-22(27-28-26)24-15-21(17(3)4)25(29)32-24/h9-10,12,16-17,20-22,24H,7-8,11,13-15H2,1-6H3/t20-,21-,22-,24-/m0/s1 |
| SMILES | COCCCc1cc(C[C@@H](C[C@H](N=[N+]=[N-])[C@@H]2C[C@@H](C(C)C)C(=O)O2)C(C)C)ccc1OC |
| IUPAC Name | (3S,5S)-5-[(1S,3S)-1-azido-3-[[4-methoxy-3-(3-methoxypropyl)phenyl]methyl]-4-methylpentyl]-3-propan-2-yloxolan-2-one |
| Molecular Weight (g/mol) | 461.59 |
| Formula Weight | 445.2941 |