CAS RN 324741-99-3
CAS RN 324741-99-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(4R)-3-[[(1S,2S)-2-Hydroxy-3-cyclopenten-1-yl]carbonyl]-4-(phenylmethyl)-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4R)-3-[[(1S,2S)-2-Hydroxy-3-cyclopenten-1-yl]carbonyl]-4-(phenylmethyl)-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.